Please login to your online account to display your discounted pricing

SB 228357, Tocris

5-HT2C/2B antagonist/inverse agonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material SB 228357
Assay Percent Range >99%
Molecular Formula C22H17F4N3O2
Formula Weight 431.39
Synonym N-[3-Fluoro-5-(3-pyrindyl)phenyl]-2, 3-dihydro-5-methoxy-6-(trifluoromethyl)-1H-indole-1-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

137510

 
r & d systems
RDS137510
10mg Each for $239.63

137550

 
r & d systems
RDS137550
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 431.39
Quantity 10mg
Chemical Name or Material SB 228357
Molecular Formula C22H17F4N3O2
CAS 181629-93-6
Assay Percent Range >99%
Synonym N-[3-Fluoro-5-(3-pyrindyl)phenyl]-2, 3-dihydro-5-methoxy-6-(trifluoromethyl)-1H-indole-1-carboxamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature