Please login to your online account to display your discounted pricing

SB 223412, Tocris™

Potent, selective non-peptide NK3 antagonist; Brain penetrant

$238.81 - $984.19

Chemical Identifiers

Chemical Name or Material SB 223412
Assay Percent Range >98%
Molecular Formula C25H22N2O2
Formula Weight 382.45
Synonym 3-Hydroxy-2-phenyl-N-[(1S)-1-phenylpropyl]-4-quinolinecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

467210

 
r & d systems
RDS467210
10mg Each for $238.81

467250

 
r & d systems
RDS467250
50mg Each for $984.19
Description & Specifications

Specifications

Formula Weight 382.45
Quantity 10mg
Chemical Name or Material SB 223412
Assay Percent Range >98%
Synonym 3-Hydroxy-2-phenyl-N-[(1S)-1-phenylpropyl]-4-quinolinecarboxamide
Molecular Formula C25H22N2O2
CAS 174636-32-9
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C