Please login to your online account to display your discounted pricing

SB 221284, Tocris

Potent, selective 5-HT2C/2B antagonist

$208.25 - $867.95

Chemical Identifiers

Chemical Name or Material SB 221284
Assay Percent Range >99%
Molecular Formula C16H14F3N3OS
Formula Weight 353.36
Synonym 2, 3-Dihydro-5-(methylthio)-N-3-pyridinyl-6-(trifluoromethyl)-1H-indole-1-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

137950

 
r & d systems
RDS137950
50mg Each for $867.95

137910

 
r & d systems
RDS137910
10mg Each for $208.25
Description & Specifications

Specifications

Formula Weight 353.36
Quantity 50mg
Molecular Formula C16H14F3N3OS
CAS 196965-14-7
Assay Percent Range >99%
Synonym 2, 3-Dihydro-5-(methylthio)-N-3-pyridinyl-6-(trifluoromethyl)-1H-indole-1-carboxamide
Chemical Name or Material SB 221284
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C