Please login to your online account to display your discounted pricing

SB 216763, Tocris

Potent, selective GSK-3 inhibitor

$106.17 - $904.70

Chemical Identifiers

Chemical Name or Material SB 216763
Assay Percent Range >98%
Molecular Formula C19H12Cl2N2O2
Formula Weight 371.22
Synonym 3-(2, 4-Dichlorophenyl)-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2, 5-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

161610

 
r & d systems
161610
10mg Each for $221.01

161650

 
r & d systems
161650
50mg Each for $904.70

16161

  SDS
r & d systems
16161
1mg Each for $106.17
Description & Specifications

Specifications

Formula Weight 371.22
Quantity 10mg
Chemical Name or Material SB 216763
Assay Percent Range >98%
Molecular Formula C19H12Cl2N2O2
Synonym 3-(2, 4-Dichlorophenyl)-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2, 5-dione
Recommended Storage Store at Room Temperature
CAS 280744-09-4
Safety and Handling

Store at Room Temperature