Please login to your online account to display your discounted pricing

SB 206553 hydrochloride, Tocris

Potent, selective 5-HT2C/5-HT2B antagonist; Orally active

$265.15 - $1,116.26

Chemical Identifiers

Chemical Name or Material SB 206553 hydrochloride
Assay Percent Range >99%
Molecular Formula C17H16N4O·HCl
Formula Weight 328.8
Synonym 3,5-Dihydro-5-methyl-N-3-pyridinylbenzo[1,2-b:4,5-b']dipyrrole-1(2H)-carboxamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

166110

 
r & d systems
RDS166110
10mg Each for $265.15

166150

 
r & d systems
RDS166150
50mg Each for $1,116.26
Description & Specifications

Specifications

Formula Weight 328.8
Quantity 10mg
CAS 1197334-04-5
Assay Percent Range >99%
Recommended Storage Desiccate at Room Temperature
Synonym 3,5-Dihydro-5-methyl-N-3-pyridinylbenzo[1,2-b:4,5-b']dipyrrole-1(2H)-carboxamide hydrochloride
Molecular Formula C17H16N4O·HCl
Chemical Name or Material SB 206553 hydrochloride
Safety and Handling

Desiccate at Room Temperature