Please login to your online account to display your discounted pricing

SB 203580, Tocris

Selective inhibitor of p38 MAPK

$133.98 - $1,154.80

Chemical Identifiers

Chemical Name or Material SB 203580
Assay Percent Range >98%
Molecular Formula C21H16FN3OS
Formula Weight 377.44
Synonym 4-[5-(4-Fluorophenyl)-2-[4-(methylsulfonyl)phenyl]-1H-imidazol-4-yl]pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

120250

 
r & d systems
RDS120250
50mg Each for $1,154.80

12021

  SDS
r & d systems
RDS12021
1mg Each for $133.98

120210

 
r & d systems
RDS120210
10mg Each for $283.27
Description & Specifications

Specifications

Formula Weight 377.44
Quantity 50mg
Assay Percent Range >98%
Chemical Name or Material SB 203580
CAS 152121-47-6
Synonym 4-[5-(4-Fluorophenyl)-2-[4-(methylsulfonyl)phenyl]-1H-imidazol-4-yl]pyridine
Molecular Formula C21H16FN3OS
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C