Please login to your online account to display your discounted pricing

SB 203186 hydrochloride, Tocris


5-HT4 antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS078550

 View more versions of this product

Catalog No. 078550

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material SB 203186 hydrochloride
Quantity 50mg
CAS 207572-69-8
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C16H20N2O2·HCl
Formula Weight 308.81
Synonym 1-Piperidinylethyl-1H-indole-3-carboxylate hydrochloride

Specifications

Chemical Name or Material SB 203186 hydrochloride
Assay Percent Range >99%
Molecular Formula C16H20N2O2·HCl
Formula Weight 308.81
Synonym 1-Piperidinylethyl-1H-indole-3-carboxylate hydrochloride
Accessories

Store at Room Temperature