Please login to your online account to display your discounted pricing

SB 203186 hydrochloride, Tocris

5-HT4 antagonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material SB 203186 hydrochloride
Assay Percent Range >99%
Molecular Formula C16H20N2O2·HCl
Formula Weight 308.81
Synonym 1-Piperidinylethyl-1H-indole-3-carboxylate hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

078510

 
r & d systems
RDS078510
10mg Each for $177.37

078550

 
r & d systems
RDS078550
50mg Each for $746.48
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 308.81
CAS 207572-69-8
Synonym 1-Piperidinylethyl-1H-indole-3-carboxylate hydrochloride
Recommended Storage Store at Room Temperature
Chemical Name or Material SB 203186 hydrochloride
Assay Percent Range >99%
Molecular Formula C16H20N2O2·HCl
Safety and Handling

Store at Room Temperature