Please login to your online account to display your discounted pricing

SB 202190, Tocris

Potent, selective inhibitor of p38 MAPK

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material SB 202190
Assay Percent Range >99%
Molecular Formula C20H14N3OF
Formula Weight 331.35
Synonym 4-[4-(4-Fluorophenyl)-5-(4-pyridinyl)-1H-imidazol-2-yl]phenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

126410

 
r & d systems
RDS126410
10mg Each for $221.01

126450

 
r & d systems
RDS126450
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 331.35
Quantity 10mg
Molecular Formula C20H14N3OF
CAS 152121-30-7
Synonym 4-[4-(4-Fluorophenyl)-5-(4-pyridinyl)-1H-imidazol-2-yl]phenol
Chemical Name or Material SB 202190
Assay Percent Range >99%
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C