Please login to your online account to display your discounted pricing

SB 200646 hydrochloride, Tocris

5-HT2C/2B antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material SB 200646 hydrochloride
Assay Percent Range >98%
Molecular Formula C15H14N4O·HCl
Formula Weight 302.76
Synonym N-(1-Methyl-1H-indol-5-yl)-N'-3-pyridinylurea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

137150

 
r & d systems
RDS137150
50mg Each for $1,050.42

137110

 
r & d systems
RDS137110
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 302.76
Quantity 50mg
Chemical Name or Material SB 200646 hydrochloride
Assay Percent Range >98%
Molecular Formula C15H14N4O·HCl
CAS 143797-62-0
Synonym N-(1-Methyl-1H-indol-5-yl)-N'-3-pyridinylurea
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature