Please login to your online account to display your discounted pricing

Thermo Scientific™ Savant™ SPD121P SpeedVac™ Kits

Conduct combinatorial chemistry applications Thermo Scientific™ SPD121P SpeedVac™ kits, which resist TFA, DSMO and other solvents.

Specifications

Chamber Material PTFE-coated Aluminum
Dimensions (L x W x H) Exterior 82 x 32 x 34 in. (2083 x 813 x 864mm)
Height (English) Exterior 34 in.
Height (Metric) Exterior 864mm
Length (English) Exterior 82 in.
View More Specs
Products
Catalog Number Mfr. No. Voltage Price Quantity    

11-716-806

 
thermo scientific™
SPD121PP1-115
115V CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chamber Material PTFE-coated Aluminum
Dimensions (L x W x H) Exterior 82 x 32 x 34 in. (2083 x 813 x 864mm)
Height (English) Exterior 34 in.
Height (Metric) Exterior 864mm
Length (English) Exterior 82 in.
Length (Metric) Exterior 2083mm
Max. Vacuum 0.1 to 20 torr endpoint shut off
Shipping Weight (English) 355 lb.
Shipping Weight (Metric) 164kg
Temperature Range (Metric) 35°C to 80°C in 5 degree increments
Width (English) Exterior 32 in.
Width (Metric) Exterior 813mm

Conduct combinatorial chemistry applications Thermo Scientific™ SPD121P SpeedVac™ kits, which resist TFA, DSMO and other solvents. All of our SpeedVac kits contain all the necessary components under one model number.

Features:

  • Deep aluminum standard PTFE-coated chamber
  • Safety cover lock
  • Radiant glass safety cover improves drying time by applying radiant IR heat to samples
  • PTFE plumbing and tubing
  • Two valve manifold with built-in automatic bleeder valve
  • Time/temperature display incorporating membrane touch pad switches
  • Variable drying times from 35° to 80°C in five degree increments
  • Manual/auto timed run
  • Endpoint mode terminates run when pre-set vacuum gauge and vacuum displays

Solvents and Solvent Combinations:

  • Acetone
  • Chloroform
  • Ethyl acetate
  • Hexane
  • Methylene chloride

Includes:

  • SpeedVac Concentrator, Enhanced Evaporation
  • -104°C, Ultra-Low Temperature Refrigerated Trap
  • OFP400 Oil-Free ETFE/PFA 4-Headed Pump
  • 64 × 1.5mL Rotor
  • Digital Vacuum Gauge
  • Chemical Trap Kit
  • Disposable Cartridge
  • Tubing and Fitting Kit
  • Glass Condensation Flask
  • CryoCool Heat Transfer Fluid (1L)

Recommended for:

  • Removing ethyl acetate from samples for CAT assay
  • Drug metabolities in solid phase extracts
  • Analyte elution solvents for evaporation