Please login to your online account to display your discounted pricing

Thermo Scientific™ Savant™ SPD111 SpeedVac™ Kits

Obtain the necessary components for molecular biology applications with Thermo Scientific™ Savant™ SPD111V P1 and P2 SpeedVac™ kits.

Specifications

Dimensions (L x W x H) Exterior 82 x 32 x 34 in. (2083 x 813 x 864mm)
Height (English) Exterior 34 in.
Height (Metric) Exterior 864mm
Length (English) Exterior 82 in.
Length (Metric) Exterior 2083mm
View More Specs
Products
Catalog Number Mfr. No. Description Electrical Requirements Includes Price Quantity    

11-716-804

 
thermo scientific™
SPD111VP1-115
SpeedVac Concentrator Kit, Basic Molecular Biology 115V 60Hz Universal Vacuum System with Oil-Free Vacuum Pump and -50C Refrigerated Vapor Trap; Glass Condensation Flask; Tubing Package, CryoCool Heat Transfer Fluid (1L) CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

11-716-805

 
thermo scientific™
SPD111VP2-115*
SpeedVac Concentrator Kit, Basic Molecular Biology 115V 60Hz -104C Refrigerated Vapor Trap; High-Vacuum Pump; 40 x 1.5mL Rotor; Universal Tubing Package; Digital Vacuum Gauge; Chemical Trap Kit; Disposable Cartridge; Glass Condensation Flask; Vacuum Pump Lubricating oil; CryoCool heat transfer fluid (1L) CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Dimensions (L x W x H) Exterior 82 x 32 x 34 in. (2083 x 813 x 864mm)
Height (English) Exterior 34 in.
Height (Metric) Exterior 864mm
Length (English) Exterior 82 in.
Length (Metric) Exterior 2083mm
Shipping Weight (English) 360 lb.
Shipping Weight (Metric) 136kg
Width (English) Exterior 32 in.
Width (Metric) Exterior 813mm

Obtain the necessary components for molecular biology applications with Thermo Scientific™ Savant™ SPD111V P1 and P2 SpeedVac™ kits. Choose our modular SPD111V P1 for standard evaporation, or the SPD111V P2 when you want to lyophillize. All of our SpeedVac kits are complete systems, containing everything you need under one part number.

Features:

  • Variable drying rate
  • Time/temperature display incorporating membrane touch pad switches
  • Safety coverlock
  • Manual/auto timed run
  • Standard, deep aluminum Teflon™ coated chamber
  • Standard glass safety lids

Solvent and Solvent Combinations:

SPD111V P1

  • Acetonitrile
  • Ethanol
  • Isopropanol
  • Methanol
  • Water
  • Acetonitrile and Water
  • Ethanol and Water
  • Methanol and Water

SPD111V P2

  • Acetonitrile
  • Ethanol
  • Methanol
  • Water
  • Acetonitrile and Water
  • Ethanol and Water
  • Methanol and Water

Includes:

SPD111V P1 SpeedVac Kit

  • SpeedVac concentrator, basic molecular biology
  • Universal vacuum system with -50°C refrigerated vapor trap
  • Glass condensation flask
  • 40 × 1.5mL rotor
  • Tubing package including 12 ft. of thick-walled vacuum tubing, cutter, clamps and fittings, BV130 manual bleed valve and instructions
  • CryoCool heat transfer fluid (1L)

SPD111V P2 SpeedVac Kit

  • SpeedVac concentrator, basic molecular biology
  • -104°C, ultra-low temperature refrigerated vapor trap
  • 40 × 1.5mL rotor
  • Universal tubing package
  • Digital vacuum gauge
  • Chemical trap kit
  • Disposable cartridge
  • Glass condensation flask
  • Vacuum pump lubricating oil
  • CryoCool heat transfer fluid (1L)

Recommended for:

  • Drying biomolecules in HPLC fractions
  • Evaporting purification solvents
  • Drying extracts of natural products in drug assays
  • Lyophillization for all the above (SPD111V P2)