Please login to your online account to display your discounted pricing

Thermo Scientific™ Savant™ ISS110 SpeedVac™ Kit

Process large batches of DNA/RNA samples efficiently with these easy to use, medium capacity kits, which contain everything you need under one part number.

Specifications

Dimensions (L x W x H) Exterior 26 x 25 x 16 in. (660 x 620 x 380mm)
Shipping Weight (English) 152 lb.
Shipping Weight (Metric) 69kg
Description ISS110 SpeedVac™ Kit
Includes Basic integrated SpeedVac system with concentrator, integated pump, -50C cold trap, 64 x 1.5mL rotor and CryoCool Heat Transfer Fluid (1L)
View More Specs
Products
Catalog Number Mfr. No. Description Electrical Requirements Price Quantity    

11-716-800

 
thermo scientific™
ISS110P1-115
ISS110 SpeedVac™ Kit 115V 50Hz CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Dimensions (L x W x H) Exterior 26 x 25 x 16 in. (660 x 620 x 380mm)
Shipping Weight (English) 152 lb.
Shipping Weight (Metric) 69kg
Description ISS110 SpeedVac™ Kit
Includes Basic integrated SpeedVac system with concentrator, integated pump, -50C cold trap, 64 x 1.5mL rotor and CryoCool Heat Transfer Fluid (1L)

Process large batches of DNA/RNA samples efficiently with the easy-to-use Thermo Scientific™ Savant™ ISS110 SpeedVac™ Kit. An ideal choice, these medium capacity kits are complete systems, containing everything you need under one part number.

Features:

  • Built-in ETFE coated vacuum pump provides years of lasting service
  • 4 liters, -50°C refrigerated vapor trap integrated
  • Choice of three drying rates

Solvents and Solvent Combinations:

  • PCR buffers (aqueous)
  • Ethanol and water
  • Isopropanol

Includes:

  • Basic integrated concentrator
  • 64× 1.5mL Rotor
  • CryoCool Heat Transfer Fluid (1L)

Recommended for:

  • DNA/RNA preparation prior to analysis by electrophoresis or microarrays
  • Labeling cDNA with fluorescent dyes
  • Drying PCR samples in aqueous buffers