Please login to your online account to display your discounted pricing

Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrator Kits

Save time and money with Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Kits. Everything you need for quick drying and concentrating of small sample volumes.

Specifications

Capacity (Metric) 4L
Certifications/Compliance CE (230V only)
Chamber Material PTFE
Displacement 31L/min. at 50Hz ; 36L/min. at 60Hz
Max. Vacuum 7 torr (9 mbar)
View More Specs
Products
Catalog Number Mfr. No. Description Voltage Price Quantity    

36-792-2

 
thermo scientific™
DNA120115
Concentrator 115V CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

20-548-133

 
thermo scientific™
DNA120OP230
DNA1200P for Oligos, 230V CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Capacity (Metric) 4L
Certifications/Compliance CE (230V only)
Chamber Material PTFE
Displacement 31L/min. at 50Hz ; 36L/min. at 60Hz
Max. Vacuum 7 torr (9 mbar)
Temperature Range (Metric) -55°C

Save time and money with compact Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Kits. These complete systems provide everything you need to quickly dry and concentrate small sample volumes in one fully integrated package, sold under one part number.

Operating Features

  • Consistent performance: designed to ensure consistent drying times and reproducible results
  • Simplicity: single program supports all DNA/RNA applications
  • User-friendly interface: with temperature/time settings
  • Three drying rates: ambient, ∽43°C, ∽65°C
  • Dual digital timers: independent control of heat/run times

Sample Protection

  • Dry at ambient temperatures or at very low heat to avoid sample denaturing
  • Oil-free diaphragm pump provides gentle transition from atmosphere to ultimate vacuum setting with no bumping

Safety Features

  • PTFE-coated chamber resists chemicals and cleans easily
  • Latched lid ensures user safety during operation
  • Unit and pump are virtually maintenance-free
  • Optional chemical traps and cartridges trap radioactivity and ammonia

Solvents and Solvent Combinations:

DNA120
  • PCR buffers (aqueous)
  • Water
  • Ethanol and water
DNA-120-OP
  • Ammonium hydroxide
  • Isopropanel
  • PCR buffers (aqueous)

ISS-110

  • PCR buffers (aqueous)
  • Ethanol and water
  • Isopropanol

DNA 120 SpeedVac Kit

  • Corrosion-resistant, PTFE coated chamber provides years of lasting service
  • Built-in, oil free vacuum pump provides maintenance-free operation
  • Integrated and compact design
  • Extremely quiet operation
  • Choice of three drying rates
  • Dual digital timers for independent control of heat and run times

DNA120OP SpeedVac Kit for oligos

  • All the features of the DNA 120
  • Plus external post trap to neutralize ammonium hydroxide, chemically resistant glass cover to contain solvents

High capacity DNA SpeedVac Kit ISS110

  • Built-in ETFE coated vacuum pump providing years of lasting service
  • Four liters, -50°C refrigerated vapor trap (integrated)
  • Choice of three drying rates

Includes:

DNA120

  • DNA concentrator with integrated pump
  • RD36 rotor for 36 × 1.6mL microtubes
DNA120-OP
  • DNA concentrator with integrated pump and glass cover
  • RD36 rotor for 36 × 1.5mL microtubes
  • ANT100 chemical trap
  • ANS121 ammonium neutralizing solution