missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ Vivaspin™ Turbo 15 PES Centrifugal Concentrators

Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius™ Vivaspin™ Turbo 15 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery. Available in a range of MWCO in 12-packs or convenient 48-pack cases.

Manufacturer:  Sartorius™ VS15T32

 View more versions of this product

Catalog No. 14-558-663


Alert:

Use Vivaspin 15 for rotors that only accept 29 x 105mm round bottom tubes (e.g. Sorvall SS34 or Beckmann JA 20).


Description

Description

Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius™ Vivaspin™ Turbo 15 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery. Available in a range of MWCO in 12-packs or convenient 48-pack cases.

Twin vertical membranes and a sleek internal design for ultra-fast concentration
  • In a single spin, 15 ml solutions can be concentrated up to 150x in 10–30 minutes
  • Large, high-flux PES membranes feature ultra-low protein binding and low extractables
  • High, constant filtration pressure allows optimal crossflow to minimize membrane fouling
Designed for maximum convenience, safety and sample recovery
  • Angular, pipette-friendly pocket facilitates easy retentate recovery of greater than 95%
  • Eliminates the risk of sample filtration to dryness
  • Stable, inert polystyrene housing with high chemical compatibility (pH 1–14)
  • UV joining technology for a smooth transition between membrane and housing
  • Volume graduation for easy monitoring of concentrate volume and 100µl dead-stop volume
Universal compatibility for rotors that accept 50mL conical bottom tubes
  • Handles sample volumes up to 15mL in swing bucket rotors (11mL in fixed angle rotors)
  • Designed for rotors that accommodate Falcon 50mL conical tubes:
    • Beckman Allegra 25R with TS-5.1-500 swing-out rotor, BUC 5 buckets, adaptors
    • Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors
    • Heraeus Multifuge 3 S-R with Sorvall 75006445 rotor, 75006441 buckets, adaptors
Specifications

Specifications

Polystyrene
Polyethersulfone
11.8cm
50,000 d
7.2cm2
11mL (Fixed Angle Rotor); 15mL (Swing Bucket Rotor)
Concentration, desalting or buffer exchange applications
4,000 x g
48/Pk.
27mm
SDS
Documents
Promotions

Promotions