missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ Vivaspin™ 500 Centrifugal Concentrators

Provides a simple, one-step concentration procedure. Sartorius™ Vivaspin™ 500 Centrifugal Concentrators are disposable ultrafiltration devices for the concentration of biological samples with starting volumes up to 500µL. Available with PES membranes in a wide range of molecular weight cut-offs.

Manufacturer:  Sartorius™ VS0121

 View more versions of this product

Catalog No. 14-558-400



Description

Description

Provides a simple, one-step concentration procedure. Sartorius™ Vivaspin™ 500 Centrifugal Concentrators are disposable ultrafiltration devices for the concentration of biological samples with starting volumes up to 500µL. Available with PES membranes in a wide range of molecular weight cut-offs.

  • Patented vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides high speed concentrations
  • Concentrate recoveries of > 90% can be reached in less than 30 minutes
  • The dead stop pocket prevents concentration to dryness for safe sample concentration
  • Universal rotor compatibility: can be used in fixed angle rotors accepting 2.2mL centrifuge tubes
  • High-flux PES membranes for ultra-fast concentration
  • Convenient concentrate recovery by pipetting
  • Clear housing with volume graduation for easy reading/monitoring
Specifications

Specifications

Yes
500μL
50mm
25/Pk.
Polycarbonate
Polyethersulfone
30,000 d
64.5mm2
SDS
Documents
Promotions

Promotions