Sartorius™ Vivacon™ 2 DNA Concentrator
Centrifugal units for fast DNA concentration and buffer exchange of 2 ml samples
Manufacturer:  Sartorius™ VN02H02
 View more versions of this product
Catalog No. 14-558-361
Description
Description
Vivacon 2 are centrifugal devices for DNA concentration and buffer exchange applications for samples up to 2 ml. For optimal performance with very diluted samples, e.g. forensic specimens, Vivacon 2 is equipped with patented Hydrosart™ regenerated cellulose membranes with extremely low unspecific binding properties and high flux. High recoveries and excellent reproducibility are paired with convenience offered by volume graduation and molecular weight cut-offs printed on individual devices. Vivacon 2 features a horizontal membrane design optimal for DNA fractionation (primer removal) as well as protein fractionation (peptide removal). The possibility of a reverse spin after sample processing assures complete concentrate recovery which is especially important when working with low sample concentrations.
  • Convenient handling in fixed-angle rotors
  • Low binding material
  • Reverse spin possibility
  • Graduation and MWCO printed on device
  • Complete and highly reproducible concentrate recovery
Specifications
Specifications
60 min.
4.92 x 0.62 in. (12.5 x 1.6cm)
4.52 in.
11.5cm
Polycarbonate Body, Polypropylene filtrate vessel, Polypropylene back spin vial, Polypropylene concentrator cap, Hydrosart™ membrane, Silicone O-ring
10000 MWCO
DNA, peptide
0.62 in.
DNA concentration, desalting, buffer exchange, fractionation
100 Pack
50ng/mL
0.95cm2
4.92 in.
12.5cm
5000xG
30 bp
10μL
1.6cm
Vivacon™ 2 DNA Concentrator
2mL