Learn More
Sartorius™ Traditional Vacuum Pumps
Neoprene membrane pumps with low noise level, oil- and maintenance-free and is a reliable vacuum
Manufacturer: Sartorius™ 16615
Description
The Microsart™ e.jet vacuum laboratory pump is designed to create sufficient vacuum for filtration and unlike traditional vacuum pumps, it allows simultaneous transfer of the filtered liquid to waste. The Microsart™ e.jet Transfer Pump is ideal for sample filtration in Microbiology achieving a trans membrane pressure below 700 mbar in compliance with ISO 8199 (Water quality - General guidance on the enumeration of micro-organisms by culture). Constant flow rates and a defined maximum vacuum guarantee smooth and reliable filtration. The Microsart™ e.jet Transfer Pump greatly reduces operating complexity and is an ideal accessory for 3-branch and 1-branch manifolds.
The Microsart™ e.jet is designed to pump both gas and liquids, meaning no loss of efficiency or malfunctions from water drawn into the pump head. The vacuum pump series Microsart™ maxi.vac and Microsart™ mini.vac provide up to date technology for daily use in the Microbiology laboratory environment. They are electric neoprene membrane pumps and therefore oil- and maintenance-free with a low noise level. The vacuum produced by these pumps is controlled and can be easily adjusted to your specifications. Thus sensitive cells (e.g. bacteria) are concentrated on the surface of a membrane filter under better conditions, which results in decreased mortality of the cells, higher recovery rates and shorter incubation times.
Specifications
110 V | |
110 V 60 Hz | |
22.5 cm | |
21.6 lb. | |
104°F | |
26 L/min. | |
60 Hz | |
25 cm | |
338 mm | |
250 mm |
13 mbar | |
13.3 x 9.8 x 8.9 in. (338 x 250 x 225 mm) | |
120 w | |
9.8 kg | |
40°C | |
Combisart | |
8.9 in. | |
13.3 in. | |
13.3 in. | |
Multiple filtration runs with multibranch manifolds for microbial enumeration |
Promotions
-
Get a Balance with $7,500 Sartorius Purchase
Buy $7,500 or more in Sartorius products and get an Entris II Precision Balance at no additional cost.: 06/30/21