Sartorius™ Sterility Testing Reusable System: Inlet Needles
For use with Sterility testing System
Manufacturer:  Sartorius™ 16964
 View more versions of this product
Catalog No. 14-555-844
Description
Description
Inlet Needle - 14-555-844
  • Stainless Steel
  • For ampoule testing, infusion bottles
  • Pierces rubber septa on side to prevent blockage by rubber particles from piercing
  • Dia. outlet hose nipple, 5mm
  • Size, long
Inlet Needle - 14-555-845
  • Stainless Steel
  • For ampoule testing, infusion bottles
  • Pierces rubber septa on side to prevent blockage by rubber particles from piercing
  • Dia. outlet hose nipple, 5mm
  • Size, short
Specifications
Specifications
5mm
Stainless Steel
Inlet Needles, 5mm, long