missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ Sterile Petri Dishes

Choose 60mm (14-555-734) or 90mm (14-555-735) diameter dishes

sterile disposable petri dishes

Manufacturer:  Sartorius™ 1431190N

 View more versions of this product

Catalog No. 14-555-735



Description

Description

 

  • Disposable
  • Type 143
  • Packaging: auto-sterile

 

Specifications

Specifications

90 mm
Sterile
Disposable
Auto-sterile
SDS
Documents
Promotions

Promotions