Sartorius Stedim Diatomaceous Earth Filter Paper
This filter grade is made of cellulose and diatomaceous earth and offer a much better separating capability than pure cellulose papers at the same rate of filtration. This grade quickly retains the finest particles at high flow rates.
Supplier: Sartorius FT4606240
Description
Applications include: clarification of beer, wine, urine during spectophotometric or refractometric tests; Filtration of the finest, semi-colloidal precipitates, e.g. those consisting of proteins, clay or cold-precipitated barium.Specifications
Disc | |
470 | |
240 mm |
Cellulose, Diatomaceous Earth | |
0.32 mm |