Sartorius™ SPH Safety Powder Hood HEPA Filters
Manufacturer:  Sartorius™ 60121
 View more versions of this product
Catalog No. 14-557-193
Description
Description
For use with Sartorius safety powder hood.
Specifications
Specifications
Filter
48 in. SPH48, SPH48B powder safety hood
99.997%
HEPA
Particulate