Please login to your online account to display your discounted pricing
Sartorius™ SENSOLUX™ EF Erlenmeyer Flasks
Specifications
Closure Type Vented
For Use With (Application) Two pre-calibrated sensor patches
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Price Quantity    
14-559-222
sartorius stedim
DCS--F2
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-559-224
sartorius™
DCS--F4
1000mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Closure Type Vented
For Use With (Application) Two pre-calibrated sensor patches
  • Single-use flask with vented cap
  • Sterile packed, ready to use
  • Patched with precalibrated pH and DO sensors (appox. 10,000 reads per sensor patch)