Sartorius Sartolab™ RF/BT Vacuum Filtration Units
Filter aqueous solutions quickly and easily. Sartorius Sartolab™ RF/BT Vacuum Filtration Units are disposable and easy to use filter units. They contain a high quality PES membrane and combine the highest flow-rates and throughput with extremely low protein binding and extractables. Available in a range of volumes.
Manufacturer:  Sartorius 180C7E
 View more versions of this product
Catalog No. 14-555-999
Description
Description
Quality, reliable sterile filtration units
  • Receiver Flask (RF) and Bottle Top (BT) units contain a high quality Sartorius PES membrane
  • Combine the highest flow-rates and throughput with extremely low protein binding and extractables
  • Ideal for filtration of tissue culture media, buffers, additives and other aqueous biologicals
  • RF and BT vacuum units are available as 150mL, 250mL, 500mL and 1000mL units
    • 500mL and 1000mL units come standard with 90mm membranes
  • Improved filtration speed and throughput for cost effective filtration
Specifications
Specifications
USP Class VI, ISO 10993
Yes
Filtration of tissue culture media, buffers, additives and other aqueous biologicals
12 Case
Polyethersulfone
0.22 μm
250 mL
Plug-on hose nipple
24 cm2
1.77 in. bottle neck
Sartolab RF Filter
Polystyrene
Sterile