Sartorius™ Quantitative Grade 393 Filter Papers
Manufacturer: Sartorius™ FT3127125
Description
Made of 100% cotton linters. Ash-free (ash content ≤ 0.01 % according to DIN 540370). Wet-strengthened. Color-coded box for easy selection.Specifications
100 per pack | |
Disc | |
Purple |
393 | |
125mm | |
0.18mm |
Promotions
-
Get a Balance with $7,500 Sartorius Purchase
Buy $7,500 or more in Sartorius products and get an Entris II Precision Balance at no additional cost.: 06/30/21