Sartorius Quantitative Grade 393 Filter Papers
These filter papers are used for quantitative and gravimetric analyses as well as for pressure or vacuum filtration. They are made out of 100 % cotton linters with an α-cellulose content of > 98 % and are acid-washed to make the papers ashless and achieve high purity.
Supplier:  Sartorius FT3127110
 View more versions of this product
Catalog No. 14-555-110
Explore available promotions
Description
Description
Quantitative Grade 393 Filter Papers
Made of 100% cotton linters. Ash-free (ash content ≤ 0.01 % according to DIN 540370). Wet-strengthened. Color-coded box for easy selection.
Specifications
Specifications
Purple
1 to 2 μm
393
0.18 mm
Disc
250 sec.
100/Pk.
110 mm