Sartorius Quantitative Grade 390 Filter Papers
Manufacturer:  Sartorius FT4103125
 View more versions of this product
Catalog No. 14-555-015
Description
Description
  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications
Specifications
Green
Folded
1 to 2μm
0.18 mm
320 mm
390
100/Pk.