Sartorius Quantitative Grade 389 Filter Papers
Manufacturer:  Sartorius FT3102100
 View more versions of this product
Catalog No. 14-555-231
Description
Description
  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications
Specifications
White
Disc
8 to 12μm
0.19 mm
100 mm
389
100/Pk.