missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ Quantitative Grade 389 F Filter Papers

$22.00 - $48.00

Specifications

Color Code Yellow
Format Disc
Grade 389
Quantity 100 per pack
Thickness 0.19mm
View More Specs

Products 4
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
14-555-114
View Documents
Sartorius™
FT3112150
150mm
Pack of 100 Each for $48.00
 
14-555-115
View Documents
Sartorius™
FT3112125
125mm
Pack of 100 Each for $34.00
 
14-555-117
View Documents
Sartorius™
FT3112090
90mm
Pack of 100 Each for $22.00
 
14-555-116
View Documents
Sartorius™
FT3112110
110mm
Pack of 100 Each for $29.00
 
Description

Description

  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications

Specifications

Yellow
389
0.19mm
Disc
100 per pack
SDS