Sartorius Quantitative Grade 292 Filter Papers
These filter papers are especially suitable for qualitative analyses that require a low ash content.
Supplier:  Sartorius FT3205090
 View more versions of this product
Catalog No. 14-555-093
Explore available promotions
Description
Description
  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications
Specifications
Disc
60 sec.
100/Pk.
90 mm
5 to 8 μm
292
0.18 mm