Sartorius Quantitative Grade 292 Filter Papers
Manufacturer:  Sartorius FT3205185
 View more versions of this product
Catalog No. 14-555-088
Description
Description
  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications
Specifications
185 mm
292
100/Pk.
Disc
5 to 8μm
0.18 mm