missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius Quantitative Grade 292 Filter Papers

$9.90 - $230.00

Specifications

Format Disc
Grade 292
Particle Retention 5 to 8μm
Quantity 100/Pk.
Thickness (Metric) 0.18 mm
View More Specs

Products 14
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
14-555-088
View Documents
Sartorius
FT3205185
185 mm
Pack of 100 Each for $40.00
 
14-555-089
View Documents
Sartorius
FT3205175
175 mm
Pack of 100 Each for $37.00
 
14-555-085
View Documents
Sartorius
FT3205320
320 mm
Pack of 100 Each for $68.00
 
14-555-086
View Documents
Sartorius
FT3205270
270 mm
Pack of 100 Each for $65.00
 
14-555-084
View Documents
Sartorius
FT3205500
500 mm
Pack of 100 Each for $230.00
 
14-555-087
View Documents
Sartorius
FT3205240
240 mm
Pack of 100 Each for $64.00
 
14-555-090
View Documents
Sartorius
FT3205150
150 mm
Pack of 100 Each for $26.00
 
14-555-091
View Documents
Sartorius
FT3205125
125 mm
Pack of 100 Each for $21.00
 
14-555-095
View Documents
Sartorius
FT3205055
55 mm
Pack of 100 Each for $12.00
 
14-555-094
View Documents
Sartorius
FT3205070
70 mm
Pack of 100 Each for $12.00
 
14-555-093
View Documents
Sartorius
FT3205090
90 mm
Pack of 100 Each for $12.00
 
14-555-092
View Documents
Sartorius
FT3205110
110 mm
Pack of 100 Each for $15.00
 
14-555-097
View Documents
Sartorius
FT3205030
30 mm
Pack of 100 Each for $9.90
 
14-555-096
View Documents
Sartorius
FT3205042
42 mm
Pack of 100 Each for $10.00
 
Description

Description

  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications

Specifications

Disc
5 to 8μm
0.18 mm
292
100/Pk.
SDS
Product Certifications