Sartorius Quantitative Grade 1289 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Supplier:  Sartorius FT3208070
 View more versions of this product
Catalog No. 14-555-062
Explore available promotions
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
Disc
100 sec.
100/Pk.
70 mm
3 to 5 μm
1290
0.21 mm