Sartorius Quantitative Grade 1289 Filter Papers
Manufacturer:  Sartorius FT3207055
 View more versions of this product
Catalog No. 14-555-071
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
55 mm
1289
100/Pk.
Disc
8 to 12μm
0.21 mm