Please login to your online account to display your discounted pricing
Sartorius™ Quantitative Grade 1289 Filter Papers
$10.00 - $51.00
Specifications
Format Disc
Quantity 100 per pack
Thickness 0.21mm
View More Specs
Products
Catalog Number Mfr. No. Grade Particle Retention Diameter (Metric) Price Quantity    
14-555-062
sartorius™
FT3208070
1290 3 to 5μmm 70mm Pack of 100 Each for $10.00
14-555-063
sartorius™
FT3208055
1290 3 to 5μmm 55mm Pack of 100 Each for $10.00
14-555-064
sartorius™
FT3207240
1289 8 to 12μmm 240mm Pack of 100 Each for $51.00
14-555-067
sartorius™
FT3207125
1289 8 to 12μmm 125mm Pack of 100 Each for $17.00
14-555-065
sartorius™
FT3207185
1289 8 to 12μmm 185mm Pack of 100 Each for $32.00
14-555-068
sartorius™
FT3207110
1289 8 to 12μmm 110mm Pack of 100 Each for $13.00
14-555-070
sartorius™
FT3207070
1289 8 to 12μmm 70mm Pack of 100 Each for $10.00
14-555-069
sartorius™
FT3207090
1289 8 to 12μmm 90mm Pack of 100 Each for $11.00
14-555-066
sartorius™
FT3207150
1289 8 to 12μmm 150mm Pack of 100 Each for $21.00
14-555-071
sartorius™
FT3207055
1289 8 to 12μmm 55mm Pack of 100 Each for $10.00
14-555-072
sartorius™
FT3207050
1289 8 to 12μmm 50mm Pack of 100 Each for $10.00
Description & Specifications
Specifications
Format Disc
Quantity 100 per pack
Thickness 0.21mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.