Sartorius Quantitative Grade 1288 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Supplier:  Sartorius FT3206047
 View more versions of this product
Catalog No. 14-555-083
Explore available promotions
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
Disc
10 sec.
100/Pk.
47 mm
12 to 15 μm
1288
0.21 mm