Sartorius Quantitative Grade 1288 Filter Papers
Manufacturer:  Sartorius FT3206090
 View more versions of this product
Catalog No. 14-555-080
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
90 mm
1288
100/Pk.
Disc
12 to 15μm
0.21 mm