Sartorius™ Printers for Secura™, Quintix™, Practum™ Analytical and Precision Weighing Balances
GLP and standard laboratory printers
$1,094.92 - $1,094.92
Specifications
For Use With (Equipment) Secura, Quintix, Practum laboratory balances
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
14-558-105 Sartorius™
YDP40
Standard Thermal Line Printer Each for $1,094.92
Specifications
Description
GLP Printer: YDP30
  • Provides GLP-compliant printouts with proper time and date stamps
  • For Secura and Quintix laboratory balances
Standard Lab Printer: YDP40
  • Transfer weight measurements to paper easily
  • Thermal printer with 203 x 203 dpi resolution
  • 120V AC adapter included
  • For Secura, Quintix and Practum laboratory balances
Description & Specifications
Specifications
Secura, Quintix, Practum laboratory balances