Sartorius™ Printer Paper/Labels and Ink for YDP Printers
Use with YDP30 and YDP40 laboratory printers. Sartorius™ Printer Paper/Labels and Ink for YDP Printers are designed to produce quick, clear printouts. Choose regular or self-adhesive paper rolls with ink ribbons for thermal transfer printing or regular paper rolls only for direct thermal printing.
Manufacturer:  Sartorius™ 69Y03287*
 View more versions of this product
Catalog No. 14-558-106
Description
Description
For use with Sartorius™ YDP30 and YDP40 laboratory printers
  • YDP30 Premium Printer prints using both direct thermal and thermal transfer printing methods
  • YDP40 Standard Printer utilizes direct thermal printing only
Specialized paper and ink ribbons
  • Direct thermal printing is ideal for less stringent requirements (paper roll only)
  • Thermal transfer printing creates fade-resistant printouts for use in regulated areas 
    • Option include both paper and ink ribbon
    • Choose between standard or self-adhesive paper rolls
Specifications
Specifications
White
Standard Lab Printer YDP40 or Premium Lab Printer YDP40
Standard Printer Paper
Set of 5