Sartorius™ Polyethersulfone Membrane Filters
Polyethersulfone (PES) filters have a high internal porosity.
Manufacturer:  Sartorius™ 1540725MIN
 View more versions of this product
Catalog No. 14-555-456
Description
Description
They perform well at high flux with an excellent throughput of aqueous solutions over the entire pH range of 1-14. Thanks to their low non-specific protein adsorption the PES type is recommended for filtering biological and pharmaceutical solutions.
Specifications
Specifications
0.2μm
25mm
Recommended for filtering biological and pharmaceutical solutions.
Polyethersulfone (PES)
100/pk.
150μm