Please login to your online account to display your discounted pricing

Sartorius™ pH Basic Benchtop Meters

Compact, basic equipment for routine measurements in the lab and reliable results in all applications

$593.52 - $936.66

Specifications

AC Adapter Yes
Accuracy (mV) ±0.02 to 0.05%mV
Accuracy (pH) ±0.005
Accuracy (Temperature) ±0.2°C
Calibration Points (pH) 3
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    

14-557-203

 
sartorius™
PB11P10.1
PY-P10 Electrode Each for $930.13

14-557-204

 
sartorius™
PB11.1
Benchtop meter only with power supply and buffer capsules Each for $593.52

14-557-803

 
sartorius™
PB11P11.1
PY-P11 TRIS compatible Glass/refillable electrode, arm and base Each for $936.66
Description & Specifications

Specifications

AC Adapter Yes
Accuracy (mV) ±0.02 to 0.05%mV
Accuracy (pH) ±0.005
Accuracy (Temperature) ±0.2°C
Calibration Points (pH) 3
Detection Range -2 to +20pH
Range (mV) ±1800mV
Range (pH) 0 to 14
Resolution (pH) 0.01
Inputs BNC
Resolution (mV) 0.1mV
Resolution (Temperature) 0.1°C
Temperature (English) Range 32° to 212°F
Temperature (Metric) Range 0° to 100°C
Type pH/mV Meter

  • Kits
    • PB-11-P20.1 Kit Includes PY-P20 electrode
    • PB-11-P10.1 Kit Includes PY-P10 electrode
    • PB-11.1 include meter only with power supply and buffer capsules

    Features

    • Automatic buffer recognition during calibration
    • Automatic temperature compensation
    • Display with stability icon next to the result
    • Simultaneous display of measured value and temperature
    • Automatic electrode check and electrode slope display during calibration
    • Easy operation with understandable symbols that practically eliminate handling errors