missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ PlatinumLINE Electrodes, pH

Get fast and accurate results with high quality materials, low-resistance A type glass membrane or unique Platinum junctions with patented Ag/AgCl reference system

$188.15 - $393.31

Specifications

Connector Types BNC
For Use With (Equipment) PY-E50-2S; PY-B1L-2S; PY-A01-2S
Reference Type Ag/AgCl
Storage Requirements Ambient temperature
Warranty 1 Year
View More Specs

Products 5
Catalog Number Mfr. No. Material Filling Solution Type Junction Price Quantity  
Catalog Number Mfr. No. Material Filling Solution Type Junction Price Quantity  
14-557-223
View Documents
Sartorius™
PYP202S
Noryl Epoxy Gel Polymer Ceramic
Each for $188.15
 
14-557-224
View Documents
Sartorius™
PYP21
Glass 3M KCl Platinum
Each for $217.92
 
14-557-225
View Documents
Sartorius™
PYP222S
Glass 3M KCl Platinum
Each for $292.33
 
14-557-227
View Documents
Sartorius™
PYP24
Glass 3M KCl Grounded Joint
Each for $393.31
 
14-558-082
View Documents
Sartorius™
PYP272S
Glass Gel polymer Hole
Each for $386.86
 
Description

Description

Electrochemical analysis is one of the most frequent and demanding tasks in daily lab routines. Measurements will differ based on the composition, temperature, conductivity and viscosity of samples and it is these different conditions that set our electrodes apart from the rest. Whether for ultrapure water, jam, wine, TRIS Buffers, or drinking water, Sartorius has the right superior and dependable electrode for your sample types and application workflows. PlatinumLINE is our most robust and diverse electrode family that offer a variety of electrode types to fit all your unique sample types with superior construction, dependable design and accurate performance.

Patented junction set sensors apart from the rest.
  • Reduce and/or eliminate metal ion contamination and chelation.
  • Ultimately minimizes damage to sensors, elongating the life of the electrodes, saving time and money.
  • Increase productivity and mitigate faulty measurements.
    • The highest quality materials result in better precision and accuracy.
    • The low resistance type A glass membranes are a perfect example of where quality materials interact with samples with less interference, yielding both speed and accuracy.
    • Liquid electrolyte electrodes with unique platinum junction and refill port slider for easier refilling and less evaporation of the electrolyte.
    • Air flush cap system prevents clogs from forming.
    • Individual serial number on each electrode for clear identification.
    • Flexibility and convenience.
    • Electrodes are available with integrated temperature sensors (ATC) or a separate temperature probe.
    • TRIS Compatibility and Impervious models available for Biological applications.
    PY-P20-2S
    Low Maintenance - pH, ceramic junction, gel filled

    PY-P21-2S
    High Performance - pH, Glass Body, Liquid 3M KCl refillable, air-flush platinum junction, TRIS compatible,break resistant conical shaped tip, ideal for biological applications, easy to clean

    PY-P22-2S
    Micro - 5mm diameter - pH, Liquid 3M KCl refillable, air flush platinum junction, TRIS,compatible ideal for low volume samples

    PY-P23-2S
    Flat-Surface - pH, Gel Filled, circular junction, ideal for meats, cheeses, agar, skin

    PY-P24-2S
    Sleeve - pH, Liquid 3M KCl refillable, Sleeve junction, ideal for low ionic samples

    PY-P27-2S
    Speartip - pH, hole junction, polymer filled
Specifications

Specifications

BNC
Ag/AgCl
1 Year
PY-E50-2S; PY-B1L-2S; PY-A01-2S
Ambient temperature
SDS