Learn More
Sartorius™ PlatinumLINE Electrodes, pH/ATC
Get fast and accurate results with high quality materials, low-resistance A type glass membrane or unique Platinum junctions with patented Ag/AgCl reference system
Manufacturer: Sartorius™ PYP12
Includes: Electrode, Manual, 50mL refill bottle
Description
Electrochemical analysis is one of the most frequent and demanding tasks in daily lab routines. Measurements will differ based on the composition, temperature, conductivity and viscosity of samples and it is these different conditions that set our electrodes apart from the rest. Whether for ultrapure water, jam, wine, TRIS Buffers, or drinking water, Sartorius has the right superior and dependable electrode for your sample types and application workflows. PlatinumLINE is our most robust and diverse electrode family that offer a variety of electrode types to fit all your unique sample types with superior construction, dependable design and accurate performance.
Patented junction set sensors apart from the rest.- Reduce and/or eliminate metal ion contamination and chelation.
- Ultimately minimizes damage to sensors, elongating the life of the electrodes, saving time and money.
- Increase productivity and mitigate faulty measurements.
- The highest quality materials result in better precision and accuracy.
- The low resistance type A glass membranes are a perfect example of where quality materials interact with samples with less interference, yielding both speed and accuracy.
- Liquid electrolyte electrodes with unique platinum junction and refill port slider for easier refilling and less evaporation of the electrolyte.
- Air flush cap system prevents clogs from forming.
- Individual serial number on each electrode for clear identification.
- Flexibility and convenience.
- Electrodes are available with integrated temperature sensors (ATC) or a separate temperature probe.
- TRIS Compatibility and Impervious models available for Biological applications.
Specifications
PY-E50-2S ; PY-B1L-2S;PY-A01-2S | |
1 Year | |
Sensing tip shape: Cylindrical with tabs | |
Ag/AgCl | |
Ambient temperature | |
NTC 10kilohm |
Noryl Epoxy | |
pH/ATC Combination Electrode | |
Ceramic | |
BNC/2.5 mm Plug | |
Gel Polymer |
Promotions
-
Get a Balance with $7,500 Sartorius Purchase
Buy $7,500 or more in Sartorius products and get an Entris II Precision Balance at no additional cost.: 06/30/21