Please login to your online account to display your discounted pricing

Sartorius™ Minisart™ SRP Syringe Filters: Sterile

Ready-to-use units for simple, rapid and reliable ultracleaning of small volume samples for the HPLC or GC analysis

$146.00 - $183.00

Specifications

Applications For simple, rapid and reliable ultracleaning of small volume samples for the HPLC or GC analysis.
Disposable Yes
Material Polytetrafluoroethylene (PTFE)
Outlet Style Luer Slip
Pore Size 0.2μm
View More Specs
Products
Catalog Number Mfr. No. Sterile Diameter (Metric) Price Quantity    

14-555-292

 
sartorius™
17573K
No 15mm Pack of 50 Each for $146.00

14-555-296

 
sartorius™
17575ACK
Yes 25mm Pack of 50 Each for $183.00
Description & Specifications

Specifications

Applications For simple, rapid and reliable ultracleaning of small volume samples for the HPLC or GC analysis.
Disposable Yes
Material Polytetrafluoroethylene (PTFE)
Outlet Style Luer Slip
Pore Size 0.2μm
Quantity 50/pk.
Type Syringe Filter

Minisart SRP units with a clean and chemically inert PTFE membrane. For solvents such as acetone, dimethylformamide and DMSO, or for aggressive aqueous liquids.

  • Sterile filtration
  • Sample cleaning
  • Particle removal
  • Easy to handle
  • Proven quality
  • 100% tested during manufacture

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.