missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius Minisart™ NML Syringe Filters, Sterile

Provides fast and reliable filtration of small to medium volumes, complete with ready-to-connect filter units. Sartorius Minisart™ NML Syringe Filters, Sterile eliminate time consuming preparatory work, such as changing out the filter and cleaning the filtration system.

Manufacturer:  Sartorius S6534FMOSK

 View more versions of this product

Catalog No. 14-555-270



Description

Description

Provides fast and reliable filtration of small to medium volumes, complete with ready-to-connect filter units. Sartorius Minisart™ NML Syringe Filters, Sterile eliminate time consuming preparatory work, such as changing out the filter and cleaning the filtration system.

Fast and reliable filtration of small to medium volumes, complete with ready-to-connect filter units eliminate time consuming preparatory work, such as changing out the filter and cleaning the filtration system. These labor saving units are used for a wide variety of applications: sample cleaning, particle removal, and sterile filtration.

  • Handy and readily available
  • Easy to use
  • Proven quality
  • Economical
  • Available in a wide array of different types
  • 100% tested during manufacture
Specifications

Specifications

Blue
200 x 130 x 100 mm
6.2 cm2
0.15 mL
Female Luer Lock
50/Pk.
50°C
Surfactant-free Cellulose Acetate
28 mm
Yes
Preparation of aqueous liquids
Meta acrylate butadiene styrene polymerisate (MBS)
Male Luer Lock
ETO-sterile
NML Syringe Filter
0.2 μm
SDS
Documents
Promotions

Promotions