missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ Minisart™ HY Syringe Filters

Designed for fast and reliable sterile venting and gas filtration. Sartorius™ Minisart™ HY Syringe Filters are single-use, complete filtration units with a naturally hydrophobic PTFE membrane and medical acrylic housing. Choose from 0.2 and 1.0µm pore sizes. Sterile option available.

$167.00 - $950.00

Specifications

Diameter (Metric) 26 mm
Material PTFE
Housing Material Medical acrylic (MBS)
Membrane Hydrophobic
Filtration Area (Metric) 5.16 cm2
View More Specs

Products 3
Catalog Number Mfr. No. Pore Size Sterility Hold up Volume (After Air Purge) Price Quantity  
Catalog Number Mfr. No. Pore Size Sterility Hold up Volume (After Air Purge) Price Quantity  
14-555-283
View Documents
Sartorius™
S6596FMOSK
0.2 μm Sterile 0.25 mL
Pack of 50 Each for $167.00
 
14-555-284
View Documents
Sartorius™
S6596FMQ
0.2 μm Non-sterile 0.25 mL
Pack of 500 Each for $950.00
 
14-555-285
View Documents
Sartorius™
1659BHYQ
1 μm Non-sterile 0.25 mL
Pack of 500 Each for $950.00
 
Description

Description

Fast and reliable sterile venting and filtration
  • Clean, safe filters that are almost free of leachables and extractables
  • Reliably removes particles and microorganisms with no leakage
  • Has minimal effect on the ingredients of the filtered solution
  • Includes a naturally hydrophobic, 26mm PTFE membrane with medical acrylic (MBS) housing
  • Wide variety of lab uses:
    • Reliable filtration of particulates and cells from gasses, additives, buffers and reagents
    • Sterile venting of small fermenters or bioreactors and of containers for culture media
    • Also used for aqueous and oily ear- or eye-solutions and other drugs
Convenient and economical
  • Complete units are ready to use
  • Single use/disposable filter units
  • Proven quality and 100% tested during manufacture
  • Sterile (via ethylene oxide) and non-sterile units available
Specifications

Specifications

26 mm
Medical acrylic (MBS)
5.16 cm2
PTFE
Hydrophobic
4.5 bar
SDS