missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ Minisart™ HY Syringe Filters

Designed for fast and reliable sterile venting and gas filtration. Sartorius™ Minisart™ HY Syringe Filters are single-use, complete filtration units with a naturally hydrophobic PTFE membrane and medical acrylic housing. Choose from 0.2 and 1.0µm pore sizes. Sterile option available.

Manufacturer:  Sartorius™ 1659BHYQ

 View more versions of this product

Catalog No. 14-555-285



Description

Description

Fast and reliable sterile venting and filtration
  • Clean, safe filters that are almost free of leachables and extractables
  • Reliably removes particles and microorganisms with no leakage
  • Has minimal effect on the ingredients of the filtered solution
  • Includes a naturally hydrophobic, 26mm PTFE membrane with medical acrylic (MBS) housing
  • Wide variety of lab uses:
    • Reliable filtration of particulates and cells from gasses, additives, buffers and reagents
    • Sterile venting of small fermenters or bioreactors and of containers for culture media
    • Also used for aqueous and oily ear- or eye-solutions and other drugs
Convenient and economical
  • Complete units are ready to use
  • Single use/disposable filter units
  • Hose barb connectors
  • Proven quality and 100% tested during manufacture
  • Sterile (via ethylene oxide) and non-sterile units available
Specifications

Specifications

Yes
PTFE
1 μm
Clear
5.16 cm2
Hose barb
4.5 bar
Cryolite
Medical acrylic (MBS)
Hydrophobic
Non-sterile
26 mm
0.25 mL
Hose barb
500/Pk.
Syringe Filter
SDS
Documents
Promotions

Promotions