Please login to your online account to display your discounted pricing
Sartorius™ Midrics Industrial Floor Scales
IP65 and IP67 protection rating suited for industrial use
$1,844.19 - $1,844.19
Products
Catalog Number Mfr. No. Price Quantity    
14-557-144
sartorius™
MW2PU1150FEL
Each for $1,844.19
Description & Specifications
There are hundreds of possible applications for an industrial scale, and not everyone needs a standard off-the-rack solution. Whatever, wherever, and however the Midrics™ industrial scale can do it all. IP65 washdown rating.
  • Painted versions have an IP65 protection rating and are therefore suited for industrial use
  • Stainless steel versions have IP67 / IP68 protection ratings and are therefore suited for industrial use in wet environments (stainless steel 1.4301/AISI 304)
  • 2 indicators (display and control unit)
  • 13 platform sizes
  • 10 weighing capacities ranging from 3 kg to 3 t
  • 2 versions for use in legal metrology of accuracy class III, resolution of 3000 e and 2 + 3000 e or standard resolution (15000 d)