missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius MGG Grade Glass Microfiber Filters w/o Binder

Binder free glass microfiber filters are suitable for analytical and gravimetric analyses. They are also used as prefilter.

$35.47 - $137.10

Specifications

Air Resistance 32mbar
Color White
Filter Type Net Filter
Format Disc
For Use With (Equipment) Filtration Systems
View More Specs

Products 10
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Price Quantity  
14-555-121
View Documents Promotion Details
Sartorius
FT31106125
0.4 in. 1.2 cm
Pack of 100 Each for $135.12
 
14-555-120
View Documents Promotion Details
Sartorius
FT31106150
5.9 in. 15 cm
Pack of 100 Each for $137.10
 
14-555-122
View Documents Promotion Details
Sartorius
FT31106110
4.33 in. 11 cm
Pack of 100 Each for $128.17
 
14-555-125
View Documents Promotion Details
Sartorius
FT31106070
2.7 in. 7 cm
Pack of 100 Each for $93.31
 
14-555-124
View Documents Promotion Details
Sartorius
FT31106090
3.5 in. 9 cm
Pack of 100 Each for $128.17
 
14-555-126
View Documents Promotion Details
Sartorius
FT31106060
2.36 in. 6 cm
Pack of 100 Each for $89.02
 
14-555-129
View Documents Promotion Details
Sartorius
FT31106047
1.8 in. 4.7 cm
Pack of 100 Each for $79.19
 
14-555-128
View Documents Promotion Details
Sartorius
FT31106050
1.96 in. 5 cm
Pack of 100 Each for $79.19
 
14-555-130
View Documents Promotion Details
Sartorius
FT31106025
0.98 in. 2.5 cm
Pack of 100 Each for $35.47
 
14-555-127
View Documents Promotion Details
Sartorius
FT31106055
2.1 in. 5.5 cm
Pack of 100 Each for $79.19
 
Description

Description

Ideal for critical analyses in which impurities can be washed out of the filter.
Specifications

Specifications

32mbar
Net Filter
Filtration Systems
1.5 μm
Non-sterile
Ideal for critical analyses in which impurities can be washed out of the filter
932°F
0.01 in.
Glass Microfiber Filter
White
Disc
Borosilicate Glass Microfiber
570 mL
38 sec.
MGG
500°C
0.28 mm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'