missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius MGF Grade Glass Microfiber Filters w/o Binder

Binder free glass microfiber filters are suitable for analytical and gravimetric analyses. They are also used as prefilter.

Supplier:  Sartorius FT31105150

 View more versions of this product

Catalog No. 14-555-134


Explore available promotions

Description

Description

Application Examples: Clarification of protein solutions, Toxicity Characteristic Leaching Procedure (TCLP) Test.
Specifications

Specifications

145mbar
Disc
0.7 μm
185 sec.
MGF
500°C
0.4 mm
15 cm
White
Borosilicate Glass Microfiber
Non-sterile
Critical analyses in which impurities can be washed out of the filter
932°F
0.015 in.
5.9 in.
SDS
Documents

Documents

Promotions

Promotions